Boron trifluoride dibutyl etherate (30 - 35% BF₃), AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Dibutyl ether-boron trifluoride, Boron trifluoride butyl etherate, Boron trifluoride butyl ether, Boron trifluoride-butyl ether complex, 1-butoxybutane- trifluoroborane(1:1), Trifluoroboron dibutyl etherate
428141000
428141000.EA
88
EUR
428141000.
Boron trifluoride dibutyl etherate (30 - 35% BF₃), AcroSeal®
Boron trifluoride dibutyl etherate
Appearance: Light yellow to brown Liquid
Formel:
BF₃·C₈H₁₈O Molecular Weight: 198,04 g/mol Dichte: 0,959 g/cm³ Flammpunkt: 70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00013195 CAS-Nummer: 593-04-4 UN: 2922 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Light yellow to brown Liquid |
Infrared spectrum | Conforms |
Strength | 30.0 to 35.0 % (Boron trifluoride) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...