Hexachloroplatinsäure Hexahydrat Pt ≥ 37,5% ≥99,95% (Metall-Basis), Premion® nitratarm

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Hydrogenhexachloroplatinat(IV) Hexahydrat, Dihydrogenhexachloroplatinat(IV) Hexahydrat

42815.03 42815.06 42815.14
42815.03EA 151 EUR
42815.03 42815.06 42815.14
Hexachloroplatinsäure Hexahydrat Pt ≥ 37,5% ≥99,95% (Metall-Basis), Premion® nitratarm
Hexachloroplatinsäure Hexahydrat

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.


Light sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.

Formel: H₂PtCl₆·6H₂O
Schmelzpunkt: 60 °C (Zers.)
Dichte: 2,430
Lagertemperatur: Raumtemperatur
MDL: MFCD00149910
CAS-Nummer: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 14,7526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR