Cobalt(II)-2-methoxyethoxide 5% (w/v) in Methylglycol , 99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
Packaged under Argon in resealable ChemSeal™ bottles
Purity: 99%
CAS No.: 142600-61-1
Molecular Formula: C6H14CoO4
Formula Weight: 209.11
Physical Form: Liquid
Sensitivity: Air and Moisture Sensitive
UN No.: UN3274
MDL No.: MFCD00798554
Formel:
C₆H₁₄CoO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798554 CAS-Nummer: 142600-61-1 UN: 1188 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...