Tribromoboranemethyl sulphide 1 M in Methylenchlorid, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

428321000
428321000.EA 135 EUR
428321000.
Tribromoboranemethyl sulphide 1 M in Methylenchlorid, AcroSeal®
Tribromoboranemethyl sulphide

Appearance: Yellow to brown Solution

Formel: C₂H₆BBr₃S
Dichte: 1,456 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00043296
CAS-Nummer: 29957-59-3
UN: 2922
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Solution
Strength 0.95 to 1.10 M
Melting point 185 to 189 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR