Methyltitanium(IV) triisopropoxide 1 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Titanium, methyltripropoxy-, Titanium,methyltris(2-propanolato)-, (T-4)-, Methyltitanium triisopropoxide, 1M in tetrahydrofuran

428371000
428371000.EA 360 EUR
428371000.
Methyltitanium(IV) triisopropoxide 1 M in THF, AcroSeal®
Methyltitanium(IV) triisopropoxide

Appearance: Yellow to orange Solution

Formel: C₁₀H₂₇O₃Ti⁻
Molecular Weight: 243,19 g/mol
Siedepunkt: 65 °C (760 mmHg)
Dichte: 0,895 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03411233
CAS-Nummer: 18006-13-8
UN: 2056
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange Solution
Strength 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR