Zinkchlorid 2 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

428561000 428568000
428561000.EA 135 EUR
428561000. 428568000.
Zinkchlorid 2 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®
Zinkchlorid
Formel: ZnCl₂
Flammpunkt: –11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011295
CAS-Nummer: 7646-85-7
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear to hazy colorless to pink to yellow-brown Solution
Water ≤0.2 %
Strength 23 to 27 %
Phosphate (PO4) ≤0.00005 %
Sulfate (SO4) ≤0.0001 %
Aluminium (Al) ≤0.000005 %
Barium (Ba) ≤0.000002 %
Bismuth (Bi) ≤0.00001 %
Cadmium (Cd) ≤0.000001 %
Calcium (Ca) ≤0.00005 %
Chromium (Cr) ≤0.000002 %
Cobalt (Co) ≤0.000001 %
Copper (Cu) ≤0.000002 %
Iron (Fe) ≤0.00002 %
Lead (Pb) ≤0.000002 %
Lithium (Li) ≤0.000001 %
Magnesium (Mg) ≤0.00001 %
Manganese (Mn) ≤0.000001 %
Molybdenum (Mo) ≤0.000002 %
Nickel (Ni) ≤0.00002 %
Potassium (K) ≤0.00001 %
Sodium (Na) ≤0.00005 %
Titanium (Ti) ≤0.00001 %
Vanadium (V) ≤0.000001 %
Zirconium (Zr) ≤0.00001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR