Silber ≥99,9% (Metall-Basis), Wolle

Lieferant: Thermo Fisher Scientific

42860.03 42860.06 42860.14
42860.03EA 33.1 EUR
42860.03 42860.06 42860.14
Silber ≥99,9% (Metall-Basis), Wolle
Silver

Purity: 99.9+% CAS No.: 7440-22-4 Physical Form: Approximately 0.05mm dia fibers Odour: Characteristic MDL No.: MFCD00003397

Formel: Ag
Siedepunkt: 2212 °C (1013 hPa)
Schmelzpunkt: 962 °C
Dichte: 10,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003397
CAS-Nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR