Multi-Element Plasma-Standardlösung 2, Specpure®

Lieferant: Thermo Fisher Scientific

42882.AE
42882.AEEA 434 EUR
42882.AE
Multi-Element Plasma-Standardlösung 2, Specpure®
Standards Kationischer Standards Mehrelement-Standards

UN Nr.: UN3264
MDL Nr.: MFCD00212338

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR