Sodium trimethylsilanolate 1 M in Dichlormethan, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Silanol, 1,1,1-trimethyl-, sodium salt (1:1), sodium trimethyl(oxido)silane, Silanol, trimethyl-, sodium salt

428841000
428841000.EA 102 EUR
428841000.
Sodium trimethylsilanolate 1 M in Dichlormethan, AcroSeal®
Sodium trimethylsilanolate
Formel: C₃H₉NaOSi
Molecular Weight: 112,179 g/mol
Dichte: 1,28 g/cm³
Flammpunkt: 63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013926
CAS-Nummer: 18027-10-6
UN: 2922
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to amber Solution (May develop some turbidity or precipitate)
Titration with HClO4 0.90 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR