Magnesium 2% in Salpetersäure 5% ≥99,9995%, TraceSELECT™ Matrixmodifizierlösung, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

428892-100ML
428892-100MLEA 355.21 EUR
428892-100ML
Magnesium 2% in Salpetersäure 5% ≥99,9995%, TraceSELECT™ Matrixmodifizierlösung, Fluka™
Magnesium

The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range.


Packaged in high-quality containers appropriate for each product
the reagents are produced and bottled under clean-room conditions.

Formel: Mg
Dichte: 1,093 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00085308
CAS-Nummer: 7439-95-4
UN: 3264
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay of Mg 1.5 - 2.5 %
Nitric acid 1.0 - 5.0 %
Metal trace analysis (ICP) Max. 5.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR