Benzylchlorformiat 50% (w/w) 3 M in Toluol, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
CBZ-Cl (Z-Cl)
428901000
428901000.EA
32
EUR
428901000.
Benzylchlorformiat 50% (w/w) 3 M in Toluol, AcroSeal®
Benzylchlorformiat
Formel:
C₈H₇ClO₂ Molecular Weight: 170,6 g/mol Dichte: 1,01 g/cm³ Flammpunkt: 7 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00000640 CAS-Nummer: 501-53-1 UN: 2920 ADR: 8,I |
Spezifikation Testergebnisse
Appearance | Clear light yellow to light pink Solution |
GC | 45 to 53 % w/w |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...