Bromcyan 3 M in Dichlormethan, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

428911000
428911000.EA 66.7 EUR
428911000.
Bromcyan 3 M in Dichlormethan, AcroSeal®
Bromcyan
Formel: CBrN
Dichte: 1,44 g/cm³
Lagertemperatur: Kühlschrank
MDL: MFCD00011597
CAS-Nummer: 506-68-3
UN: 2927
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Solution
Strength 2.95 to 3.30 M
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR