Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in Toluol, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

428981000
428981000.EA 411 EUR
428981000.
Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in Toluol, AcroSeal®
Bis(2-methoxyethyl)aminosulphur trifluoride
Formel: C₆H₁₄F₃NO₂S
Molecular Weight: 221.24 g/mol
Dichte: 1.2 g/cm³
Flammpunkt: 4 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01321415
CAS-Nummer: 202289-38-1
UN: 3286
ADR: 3 II

Bestellen

Spezifikation Testergebnisse

Appearance Dark yellow to orange Liquid
NMR ≥95 % (On neat substance)
Titration with HClO4 ≥50 %
GC ≥97.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR