Lithiumtetrachlorocuprat 0.1M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Dilithium tetrachlorocuprate
429051000
429051000.EA
104
EUR
429051000.
Lithiumtetrachlorocuprat 0.1M in THF, AcroSeal®
Lithiumtetrachlorocuprat
Formel:
Cl₄CuLi₂ Molecular Weight: 219.24 g/mol Dichte: 0.91 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011081 CAS-Nummer: 15489-27-7 UN: 1993 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear orange to red-brown Solution |
Titration Argentometric | 0.09 to 0.12 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...