Triethyloxoniumtetrafluoroborat 1M in Methylenchlorid, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

429161000
429161000.EA 126 EUR
429161000.
Triethyloxoniumtetrafluoroborat 1M in Methylenchlorid, AcroSeal®
Triethyloxonium tetrafluoroborate
Formel: C₆H₁₅BF₄O
Molecular Weight: 189,99 g/mol
Dichte: 1,328 g/cm³
Lagertemperatur: Gefrierschrank
MDL: MFCD00044423
CAS-Nummer: 368-39-8
UN: 2922
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear to slightly hazy colorless to yellow to pink to brown Solution (May develop some turbidity or precipitate)
Titration with NaOH 0.90 to 1.10 M
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR