Kupfer(II)sulfid ≥99,8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
42925.18
42925.30
42925.A1
42925.18EA
109
EUR
42925.18
42925.30
42925.A1
Kupfer(II)sulfid ≥99,8% (Metall-Basis)
Kupfer(II)sulfid
Purity: 99.8%
CAS No.: 1317-40-4
Molecular Formula: CuS
Formula Weight: 95.60
Physical Form: -200 Mesh Powder
Density: 3.9-4.6
Refractive Index: 1.45
Sensitivity: Air and Moisture Sensitive
Melting Point: 220° dec.
Merck Reference: 14,2655
MDL No.: MFCD00016066
Formel:
CuS Schmelzpunkt: 220 °C (Zers.) Dichte: 3,9…4.6 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016066 CAS-Nummer: 1317-40-4 EINECS: 215-271-2 Merck Index: 14,2655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...