Chrom ≥99,999% (Metall-Basis), Granulat -4/+7 mesh
Lieferant: Thermo Fisher Scientific
42926.18
42926.18EA
240
EUR
42926.18
Chrom ≥99,999% (Metall-Basis), Granulat -4/+7 mesh
Chromium
Purity: 99.999%
CAS No.: 7440-47-3
Odour: Odourless
MDL No.: MFCD00010944
Formel:
Cr Siedepunkt: 2469 °C (1013 hPa) Schmelzpunkt: 1840 °C Dichte: 7,2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010944 CAS-Nummer: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...