Triethylorthoformiat, wasserfrei 98%, AcroSeal®

Lieferant: Thermo Fisher Scientific
Achtung

429271000 429278000
429271000.EA 65.4 EUR
429271000. 429278000.
Triethylorthoformiat, wasserfrei 98%, AcroSeal®
Triethylorthoformiat
Formel: C₇H₁₆O₃
Siedepunkt: 146 °C (760 mmHg)
Schmelzpunkt: –61 °C
Dichte: 0,89 g/cm³
Flammpunkt: 35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009230
CAS-Nummer: 122-51-0
UN: 2524
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥97.5 %
Residue after evaporation ≤0.005 %
Acidity ≤0.0001 meq/g
Substance darkened by H2SO4 ≤10 APHA
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.2 ppm
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.001 %
Bromine number ≤0.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR