Germanium ≥99,9999% (Metall-Basis), polykristalline Stangen, Ø 6.35 mm (0.25 in)
Lieferant: Thermo Fisher Scientific
42951.BS
42951.BSEA
637
EUR
42951.BS
Germanium ≥99,9999% (Metall-Basis), polykristalline Stangen, Ø 6.35 mm (0.25 in)
Germanium
Purity: 99.9999%
CAS No.: 7440-56-4
Physical Form: ≈1.7g/cm
Odour: Odourless
MDL No.: MFCD00085310
Formel:
Ge Siedepunkt: 2830 °C (1013 hPa) Schmelzpunkt: 937,2 °C Dichte: 5,323 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085310 CAS-Nummer: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...