Graphit ≥99,8% (Metall-Basis), Folie, Dicke 0.4 mm (0.015 in)

Lieferant: Thermo Fisher Scientific

42952.CG 42952.VA 42952.VU
42952.CGEA 126 EUR
42952.CG 42952.VA 42952.VU
Graphit ≥99,8% (Metall-Basis), Folie, Dicke 0.4 mm (0.015 in)
Graphit

Purity: 99.8% MDL No.: MFCD00144065

Formel: C
Molecular Weight: 12.01 g/mol
Schmelzpunkt: 3827
Dichte: 2.09…2.23 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR