Propionaldehyde 2,4-dinitrophenylhydrazone standard_analytical, environmental analysis, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

429546-20MG
429546-20MGEA 405 EUR
429546-20MG
Propionaldehyde 2,4-dinitrophenylhydrazone standard_analytical, environmental analysis, Sigma-Aldrich®
Propionaldehyde 2,4-dinitrophenylhydrazone
Formel: C₉H₁₀N₄O₄
Molecular Weight: 238.20 g/mol
Schmelzpunkt: 152…155 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191484
CAS-Nummer: 725-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06cb9e510a6f7723b6eb114ce1cbbf55688d4666&vid=009e9b3b273a0116925fd32388f83dec9cf94e91&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR