Acetaldehyde 2,4-dinitrophenylhydrazone standard_analytical, environmental analysis, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Acetaldehyde 2,4-Dinitrophenylhydrazone

429554-20MG
429554-20MGEA 405 EUR
429554-20MG
Acetaldehyde 2,4-dinitrophenylhydrazone standard_analytical, environmental analysis, Sigma-Aldrich®
Acetaldehyde 2,4-dinitrophenylhydrazone
Formel: C₈H₈N₄O₄
Molecular Weight: 224.18 g/mol
Schmelzpunkt: 165…168 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191298
CAS-Nummer: 1019-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f0f95a4c7674ba319560356c616d3ea64fdf2b1&vid=6874a585ce1ac0fa5139f0b98d2be81eaf0996ff&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR