Boron trifluoridedimethyl sulphide complex, purified

Lieferant: Thermo Fisher Scientific
Gefahr


Argon_Chemseal

42970.AE
42970.AEEA 298 EUR
42970.AE
Boron trifluoridedimethyl sulphide complex, purified
Boron trifluoridedimethyl sulphide complex

Packaged under Argon in resealable ChemSeal™ bottles


CAS No.: 353-43-5 Molecular Formula: BF3.S(CH3)2 Formula Weight: 129.94 Physical Form: Liquid Density: 1.235 Sensitivity: Moisture Sensitive UN No.: UN2920 MDL No.: MFCD00013193

Formel: BF₃·S(CH₃)₂
Molecular Weight: 129.94 g/mol
Dichte: 1.235
Lagertemperatur: Raumtemperatur
MDL: MFCD00013193
CAS-Nummer: 353-43-5
UN: 2920
ADR: 8 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR