Blei ≥99,999% (Metall-Basis), Barren
Lieferant: Thermo Scientific
|
Gefahr
|
43008.22
43008.22EA
172
EUR
43008.22
Blei ≥99,999% (Metall-Basis), Barren
Lead
Purity: 99.999%
CAS No.: 7439-92-1
Appearance: Grey
Odour: Odourless
MDL No.: MFCD00134050
Formel:
Pb Molecular Weight: 207,2 g/mol Siedepunkt: 1740 °C (1013 hPa) Schmelzpunkt: 327 °C Dichte: 11,3437 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134050 CAS-Nummer: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...