Tetraethylammoniumhydroxid 35% (w/w) in wässriger Lösung, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 77-98-5
Molecular Formula: C8H21NO
Formula Weight: 147.26
Physical Form: Liquid
Density: 1.023
Refractive Index: 1.4040
Sensitivity: Air Sensitive
Odour: Amine-like
Boiling Point: dec.
Beilstein Reference: 3912961
Merck Reference: 14,9201
UN No.: 3267
MDL No.: MFCD00009024
Formel:
C₈H₂₁NO Siedepunkt: Zersetzung Dichte: 1,023 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009024 CAS-Nummer: 77-98-5 EINECS: 201-073-3 UN: 3267 ADR: 8,II Merck Index: 14,9201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...