Zirkonium ≥99,9% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)

Lieferant: Thermo Fisher Scientific

43049.FQ 43049.FF 43049.FW
43049.FQEA 183 EUR
43049.FQ 43049.FF 43049.FW
Zirkonium ≥99,9% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)
Zirkonium

Purity: 99.9+% CAS No.: 7440-67-7 Physical Form: ≈0.1g/25x25mm Odour: Odourless MDL No.: MFCD00011303

Formel: Zr
Lagertemperatur: Raumtemperatur
MDL: MFCD00011303
CAS-Nummer: 7440-67-7
EINECS: 231-176-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR