Succinate standard for IC, TraceCERT®, Supelco®
Lieferant: Merck
43057-100ML
43057-100MLEA
58.2
EUR
43057-100ML
Succinate standard for IC, TraceCERT®, Supelco®
Standards
Anionische Standards
Succinate-Standards
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.