Siliciumdioxid in Ethylenglykol, kolloidale Dispersion

Lieferant: Thermo Scientific

Synonyme: Silicium(IV)oxid

43091.A1
43091.A1EA 54.3 EUR
43091.A1
Siliciumdioxid in Ethylenglykol, kolloidale Dispersion
Siliciumdioxid

CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: 0.02 Micron Particles, in Liquid Density: 1.3 Appearance: White Odour: Sweet Solubility: Miscible in water. Partially miscible in alcohols, ketones. Immiscible in hydrocarbons. MDL No.: MFCD00011232

Formel: SiO₂
Molecular Weight: 60,08 g/mol
Dichte: 1,3
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 7631-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR