Siliciumdioxid 30% in Ethylenglykol, kolloidale Dispersion
Lieferant: Thermo Fisher Scientific
Synonyme:
Silicium(IV)oxid
43093.A1
43093.A1EA
84.1
EUR
43093.A1
Siliciumdioxid 30% in Ethylenglykol, kolloidale Dispersion
Siliciumdioxid
CAS No.: 7631-86-9
Molecular Formula: O2Si
Formula Weight: 60.09
Physical Form: 0.05 Micron Particles, in Liquid
Density: 1.3
Appearance: White
Odour: Sweet
MDL No.: MFCD00011232
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 Schmelzpunkt: 1719 Dichte: 1.3 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 7631-86-9 Merck Index: 14,08493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...