Siliciumdioxid 30% in Ethylenglykol, kolloidale Dispersion

Lieferant: Thermo Scientific

Synonyme: Silicium(IV)oxid

43094.A1
43094.A1EA 80 EUR
43094.A1
Siliciumdioxid 30% in Ethylenglykol, kolloidale Dispersion
Siliciumdioxid

CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 0.1 Micron Particles, in Liquid Appearance: White Odour: Sweet MDL No.: MFCD00011232

Formel: SiO₂
Molecular Weight: 60,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 7631-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR