Ammoniumacetat 99,99
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Essigsäure Ammoniumsalz
431311-250G
431311-50G
431311-250GEA
469.98
EUR
431311-250G
431311-50G
Ammoniumacetat 99,99
Ammoniumacetat
Formel:
H₃CCOONH₄ Molecular Weight: 77.08 g/mol Schmelzpunkt: 113 °C Dichte: 1.170 g/cm³ (20 °C) Flammpunkt: 136 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013066 CAS-Nummer: 631-61-8 EINECS: 211-162-9 Merck Index: 13,00495 |
|
Spezifikation Testergebnisse
Appearance of the substance | complying |
Identity (IR) | complying |
Assay | Min. 97.0 % |
pH (5 %, 25 °C) | 6.7 - 7.3 |
Sulfated ash | Max. 0.01 % |
Water insoluble matter | Max. 0.005 % |
Metal trace analysis (ICP) | Max. 100 ppm |
Iron (Fe) | Max. 5 ppm |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 5 ppm |
Nitrate (NO₃) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...