trans-4-Octen ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 14850-23-8
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 21°(70°F)
Physical Form: Liquid
Density: 0.715
Refractive Index: 1.4120
Odour: Characteristic
Boiling Point: 122-123°
Beilstein Reference: 1719104
UN No.: 3295
MDL No.: MFCD00009476
Formel:
C₈H₁₆ Siedepunkt: 122…123 °C Dichte: 0,715 Flammpunkt: 21 °C (70 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009476 CAS-Nummer: 14850-23-8 EINECS: 238-913-3 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...