Kaliumhydrogencarbonat ≥99,95% (99.7-100.5% dry basis)

Lieferant: Honeywell Chemicals

Synonyme: Kaliumbicarbonat

431583-250G
431583-250GEA 460.47 EUR
431583-250G
Kaliumhydrogencarbonat ≥99,95% (99.7-100.5% dry basis)
Kaliumhydrogencarbonat
Formel: KHCO₃
Molecular Weight: 100.12 g/mol
Schmelzpunkt: 100…120 °C (Zers.)
Dichte: 2.17 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011402
CAS-Nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay (excluding dried substances) 99.7 - 100.5 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 5.0 ppm
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.03 %
Metal trace analysis (ICP) Max. 500.0 ppm
Heavy metals (as Pb) Max. 5 ppm
S-compounds (as SO₄) Max. 0.003 %
Ammonium (NH₄) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR