3-Bromobenzylmagnesium bromide 0.25 M in Diethylether, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
431601000
431601000.EA
286
EUR
431601000.
3-Bromobenzylmagnesium bromide 0.25 M in Diethylether, AcroSeal®
3-Bromobenzylmagnesium bromide
Formel:
C₇H₆Br₂Mg Molecular Weight: 274,238 g/mol Dichte: 0,766 g/cm³ Flammpunkt: –40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319902 CAS-Nummer: 107549-22-4 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear yellow Solution |
Titration Grignard | 0.15 to 0.35 M |
Titration with HClO4 | ≥90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...