[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
431650500
431650500.EA
241
EUR
431650500.
[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide
Formel:
C₁₁H₁₄BrMgN Molecular Weight: 264,449 g/mol Dichte: 0,919 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840891 CAS-Nummer: 480424-80-4 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear brown Solution |
Titration Grignard | 0.15 to 0.35 M |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...