Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N,N'-Disalicylalethylenediamine Cobalt(II), Salcomine, N,N'-Bis(salicylidene)ethylenediiminocobalt(II), N,N'-Ethylenebis(salicylideneiminato)cobalt(II)

43174.22 43174.03 43174.14 43174.06
43174.22EA 506 EUR
43174.22 43174.03 43174.14 43174.06
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)]

Purity: 96% CAS No.: 14167-18-1 Molecular Formula: C16H14CoN2O2 Formula Weight: 325.23 Physical Form: Powder Appearance: Black Odour: Odourless MDL No.: MFCD00000009

Formel: C₁₆H₁₆CoN₂O₂
Molecular Weight: 327.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000009
CAS-Nummer: 14167-18-1
EINECS: 238-012-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR