[2-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
431750500
431750500.EA
249
EUR
431750500.
[2-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
[2-(4-Morpholinylmethyl)phenyl]magnesium bromide
Formel:
C₁₁H₁₄BrMgNO Dichte: 0,926 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840888 CAS-Nummer: 480424-77-9 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear yellow Solution |
Titration Grignard | 0.15 to 0.35 M |
Titration with NaOH | ≥98.5 % |
Chloride (Cl) | ≤5 ppm |
Phosphate (PO4) | ≤5 ppm |
Calcium (Ca) | ≤10 ppm |
Copper (Cu) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Potassium (K) | ≤10 ppm |
Sodium (Na) | ≤20 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...