[4-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
431770500
431770500.EA
315
EUR
431770500.
[4-(4-Morpholinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
[4-(4-Morpholinylmethyl)phenyl]magnesium bromide
Formel:
C₁₁H₁₄BrMgNO Dichte: 0,926 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840890 CAS-Nummer: 480424-75-7 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear yellow Solution |
Titration Grignard | 0.15 to 0.35 M |
Chloride (Cl) | ≤0.05 % |
Phosphate (PO4) | ≤0.03 % |
Sulfate (SO4) | ≤0.05 % |
Iron (Fe) | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...