Certrinitrat hydrat ≥99,998% (Metall-Basis), REacton®

Lieferant: Thermo Fisher Scientific
Gefahr

43181.22 43181.14
43181.22EA 521 EUR
43181.22 43181.14
Certrinitrat hydrat ≥99,998% (Metall-Basis), REacton®
Certrinitrat

Purity: 99.998% Molecular Formula: Ce(NO3)3.xH2O (x~6-7) Formula Weight: 326.13(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Odour: Acidic Merck Reference: 14,2000 UN No.: 1477 MDL No.: MFCD00010926

Formel: Ce(NO₃)₃·xH₂O
Siedepunkt: 200 °C (1013 hPa)
Schmelzpunkt: 39 °C
Dichte: 2,377 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010926
CAS-Nummer: 10108-73-3
EINECS: 233-297-2
UN: 1477
ADR: 5.1,III
Merck Index: 14,2000

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR