3-Methoxybenzylmagnesium chloride 0.25 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
431820500
431820500.EA
147
EUR
431820500.
3-Methoxybenzylmagnesium chloride 0.25 M in THF, AcroSeal®
3-Methoxybenzylmagnesium chloride
Formel:
C₈H₉ClMgO Dichte: 0,904 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319896 CAS-Nummer: 26905-40-8 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear yellow to grey to brown Solution |
Titration Grignard | 0.15 to 0.35 M |
Particle size | (D50) 205 µm (Typical) |
Aluminium (Al) | ≤250 ppm (Typical) |
Calcium (Ca) | ≤80 ppm (Typical) |
Chromium (Cr) | ≤0.5 ppm (Typical) |
Iron (Fe) | ≤65 ppm (Typical) |
Potassium (K) | ≤50 ppm (Typical) |
Titanium (Ti) | ≤150 ppm (Typical) |
Silicon dioxide (SiO2) | ≥99.80 % (Typical) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...