3-Fluoro-4-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
bromo(3-fluoro-4-methoxyphenyl)magnesium, Magnesium,bromo(3-fluoro-4-methoxyphenyl)-
431870500
431870500.EA
101
EUR
431870500.
3-Fluoro-4-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
3-Fluoro-4-methoxyphenylmagnesium bromide
Formel:
C₇H₆BrFMgO Molecular Weight: 229,331 g/mol Dichte: 0,964 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311480 CAS-Nummer: 112780-67-3 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear yellow to brown Solution |
Titration Grignard | 0.40 to 0.60 M |
HPLC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...