5-Fluoro-2-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Magnesium,bromo(5-fluoro-2-methoxyphenyl)-, bromo(5-fluoro-2-methoxyphenyl)magnesium

431880500
431880500.EA 94.9 EUR
431880500.
5-Fluoro-2-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
5-Fluoro-2-methoxyphenylmagnesium bromide
Formel: C₇H₆BrFMgO
Molecular Weight: 229,331 g/mol
Dichte: 0,948 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01311481
CAS-Nummer: 188132-02-7
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
Melting point 67 to 70 °C
GC ≥99.0 %
Enantiomeric excess ≥99.0 %
Specific optical rotation +53° to +59° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR