(1,3-Dioxan-2-ylethyl)magnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Magnesium, bromo[2-(1,3-dioxan-2-yl)ethyl]-, Bromo[2-(1,3-dioxan-2-yl)ethyl]magnesium

431900500
431900500.EA 136 EUR
431900500.
(1,3-Dioxan-2-ylethyl)magnesium bromide 0.5 M in THF, AcroSeal®
(1,3-Dioxan-2-ylethyl)magnesium bromide
Formel: C₆H₁₁BrMgO₂
Dichte: 0,951 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061518
CAS-Nummer: 78078-50-9
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
Melting point 64 to 70 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR