3,5-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

431940500
431940500.EA 202 EUR
431940500.
3,5-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,5-Dimethylphenylmagnesium bromide
Formel: C₈H₉BrMg
Molecular Weight: 209.37 g/mol
Dichte: 0.91 g/cm³
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01311497
CAS-Nummer: 34696-73-6
UN: 2924
ADR: 3 II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Solution
Titration Grignard 0.40 to 0.60 M
Melting point 96 to 99 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR