3,5-Dimethyl-4-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Scientific
|
Gefahr
|
431960500
431960500.EA
104
EUR
431960500.
3,5-Dimethyl-4-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,5-Dimethyl-4-methoxyphenylmagnesium bromide
Formel:
C₉H₁₁BrMgO Molecular Weight: 239,395 g/mol Dichte: 0,949 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319917 CAS-Nummer: 185416-17-5 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Yellow to brown Solution |
Titration Grignard | 0.40 to 0.60 M |
Refractive index | 1.5830 to 1.5850 (20°C, 589 nm) |
GC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...