Pestle for micro-tissue grinders with screw caps
Lieferant: DWK Life Sciences
357537
432-5004EA 216 EUR
432-5004
Pestle for micro-tissue grinders with screw caps
Mörser und Pistille Pistille
Replacement pestle for WHEATON® micro tissue grinder with cap, nominal capacity 0,5 ml.
  • Manufactured from borosilicate glass 3.3
  • With PTFE pestle and screw cap
  • 13-425 open top screw cap and PTFE faced silicone rubber liner
  • Autoclavable