Tetrabutylammoniumchlorid Hydrat 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
432060050
432060250
432061000
432060050.EA
25.1
EUR
432060050.
432060250.
432061000.
Tetrabutylammoniumchlorid Hydrat 96%
Tetrabutylammoniumchlorid Hydrat
Formel:
C₁₆H₃₆ClN·xH₂O Schmelzpunkt: 37…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011635 CAS-Nummer: 37451-68-6 |
Spezifikation Testergebnisse
Appearance | Off-white Low melting solid |
Infrared spectrum | Conforms |
Titration Argentometric | ≥95.0 % (On dried substance) |
Water | ≤10.0 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...