Triethylamin 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

432280010 432280025
432280010.EA 66.5 EUR
432280010. 432280025.
Triethylamin 99.5% zur Analyse
Triethylamin
Formel: (C₂H₅)₃N
Siedepunkt: 90 °C (760 mmHg)
Schmelzpunkt: –115 °C
Dichte: 0,728 g/cm³
Flammpunkt: –11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009051
CAS-Nummer: 121-44-8
UN: 1296
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20°C, 589 nm)
Water ≤0.1 %
GC ≤0.05 % (ethanol)
Color scale ≤15 APHA
Residue after evaporation ≤0.002 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR