Tris(dimethylamino)aluminum dimer
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
unter Argon abgefüllt
CAS No.: 32093-39-3
Molecular Formula: C12H36Al2N6
Formula Weight: 318.42
Flash Point: 21°(70°F)
Physical Form: Powder, packaged under argon
Density: 0.865
Sensitivity: Moisture Sensitive
Appearance: White to pale brown
Melting Point: 87-89°
Boiling Point: 90°/0.05mm subl.
UN No.: UN3131
MDL No.: MFCD00269811
Applications: Preparation of aluminum nitrides
Formel:
C₁₂H₃₆Al₂N₆ Siedepunkt: 90 °C (0,05 mmHg) (Subl.) Schmelzpunkt: 87…89 °C Dichte: 0,865 Flammpunkt: 21 °C (70 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269811 CAS-Nummer: 32093-39-3 UN: 3131 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...