2-Cyan-5-methylanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Cyano-5-methylaniline

43260.03 43260.14
43260.03EA 51.5 EUR
43260.03 43260.14
2-Cyan-5-methylanilin ≥98%
2-Cyan-5-methylanilin

Purity: 98% CAS No.: 26830-96-6 Molecular Formula: C8H8N2 Formula Weight: 132.17 Physical Form: Powder Melting Point: 91-93° UN No.: 3439 MDL No.: MFCD00173706

Formel: C₈H₈N₂
Molecular Weight: 132.17
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173706
CAS-Nummer: 26830-96-6
EINECS: 248-020-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR